missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 4-(aminocarbothioyl)tetrahydropyridine-1(2H)-carboxylate, 95+%, Thermo Scientific™

Catalog Number 10426302
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
214834-18-1
C11H20N2O2S
244.353
SCGQNJHAAYUQOO-UHFFFAOYSA-N
1-boc-4-thiocarbamoylpiperidine, tert-butyl 4-aminocarbothioyl tetrahydropyridine-1 2h-carboxylate, 1-tert-butoxycarbonylpiperidine-4-carbothioamide, tert-butyl 4-thiocarbamoylpiperidine-1-carboxylate, tert-butyl 4-aminocarbonothioyl piperidine-1-carboxylate, 4-thiocarbamoyl-piperidine-1-carboxylic acid tert-butyl ester, piperidine-4-thiocarboxamide, n1-boc protected, 1-tert-butoxycarbonyl-4-carbamothioylpiperidine, 1-tert-butoxycarbonyl piperidine-4-thiocarboxamide, tert-butyl 4-thiocarbamoyl-piperidine-1-carboxylate
2735648
tert-butyl 4-carbamothioylpiperidine-1-carboxylate
CC(C)(C)OC(=O)N1CCC(CC1)C(=S)N
This item is not returnable. View return policy
214834-18-1
C11H20N2O2S
244.353
SCGQNJHAAYUQOO-UHFFFAOYSA-N
1-boc-4-thiocarbamoylpiperidine, tert-butyl 4-aminocarbothioyl tetrahydropyridine-1 2h-carboxylate, 1-tert-butoxycarbonylpiperidine-4-carbothioamide, tert-butyl 4-thiocarbamoylpiperidine-1-carboxylate, tert-butyl 4-aminocarbonothioyl piperidine-1-carboxylate, 4-thiocarbamoyl-piperidine-1-carboxylic acid tert-butyl ester, piperidine-4-thiocarboxamide, n1-boc protected, 1-tert-butoxycarbonyl-4-carbamothioylpiperidine, 1-tert-butoxycarbonyl piperidine-4-thiocarboxamide, tert-butyl 4-thiocarbamoyl-piperidine-1-carboxylate
2735648
tert-butyl 4-carbamothioylpiperidine-1-carboxylate
CC(C)(C)OC(=O)N1CCC(CC1)C(=S)N
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 244.36
Percent Purity 95%
Chemical Name or Material tert-Butyl 4-(aminocarbothioyl)tetrahydropyridine-1(2H)-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'