missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl N-(3-formylbenzyl)carbamate, 90%, Thermo Scientific™

Catalog Number p-4510274
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
170853-04-0
C13H17NO3
235.283
JORXNWZTJLYRQA-UHFFFAOYSA-N
2794834
tert-butyl N-[(3-formylphenyl)methyl]carbamate
CC(C)(C)OC(=O)NCC1=CC=CC(=C1)C=O
This item is not returnable. View return policy
170853-04-0
C13H17NO3
235.283
JORXNWZTJLYRQA-UHFFFAOYSA-N
2794834
tert-butyl N-[(3-formylphenyl)methyl]carbamate
CC(C)(C)OC(=O)NCC1=CC=CC(=C1)C=O
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym tert-butyl 3-formylbenzylcarbamate, tert-butyl n-3-formylphenyl methyl carbamate, tert-butyl 3-formylbenzyl carbamate, 3-aminomethyl benzaldehyde, n-boc protected, 3-formylbenzylamine, n-boc protected, tert-butyl n-3 formylbenzyl carbamate, tert-butyl n-3-formylbenzyl carbamate, tert-butyl3-formylbenzylcarbamate, 3-formylbenzyl carbamic acid tert-butyl ester, 3-formyl-benzyl-carbamic acid tert-butyl ester
Formula Weight 235.28
Percent Purity 90%
Chemical Name or Material tert-Butyl 3-formylbenzylcarbamate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'