missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Test Tube Caps

Reduce risk of sample contamination and spillage with a polyethylene constructed cap. Kartell™ Test Tube Caps are specially designed to accommodate both platic and glass test tubes.

Brand:  Kartell™ 0065500

Code : A0

Additional Details : Weight : 0.09000kg

 View more versions of this product

Product Code. 10737721

  • 9.50 € / Pack of 100
Estimated Shipment: 13-06-2024
to see stock.
Add to basket

Description

Description

  • Constructed of PE
  • Designed to fit plastic or glass test tubes
Specifications

Specifications

CE/IVD
Polyethylene
24 mm Diameter Test Tubes
22 mm
Natural
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers