Learn More
Thermo Scientific™ Test Tube Racks for MaxQ™ Shaker
Mix and match these shaker test tube racks, available in a variety of sizes and arrays, to suit individual needs in the laboratory.
Brand:  Thermo Scientific™ 30180BI
Code : 50
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 10174383
  • 364.00 € / Each
In Stock
to see stock.
Add to basket
Description
Description
Mix and match Thermo Scientific™ MaxQ™ Shaker Test Tube Racks, available in a variety of sizes and arrays, to suit individual needs in the laboratory. All racks are compatible with MaxQ shaker platforms.
Racks are available in a multitude of sizes, arrays, and colors. Compatible with MaxQ shaker platforms.
Specifications
Specifications
Test Tube Rack
6 x 12
10 to 13 mm Tubes
MaxQ™ Shaker Platform
Red; 10 to 13 mm; 6 x 12 array
Red
72