Learn More
Grant Instruments™ Test Tube Tray for Grant Shaking Water Baths
Holds 3 to 5 SR Racks. For use with Grant Shaking Water Baths
Brand: Grant Instruments™ TS12
Code : NEW
Additional Details : Weight : 2.00000kg
Description
Include threaded holes to accept flask clamps or holder for deep well plates (=2ml). Grant Instruemnts™ Flask Tray for Grant Shaking Water Baths are compatible with SR plate holders and racks or can be used alone to accommodate bags and miscellaneous vessels.
Test Tube Tray for Grant Shaking Water Baths
- TS12: Holds up to 3 SR Racks for LSB12 Model Aqua Pro Shaking Water Bath
- TS18: Holds up to 5 SR Racks for LSB18 Model Aqua Pro Shaking Water Bath
- TS26: Holds up to 5 SR Racks for OLS26 Model Aqua Pro Shaking Water Bath
Specifications
LSB12 | |
Test Tube Tray | |
Up to 5 SR Racks | |
Stainless Steel |