missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium hydroxide, 1M solution in methanol, Thermo Scientific Chemicals

Brand:  Thermo Scientific Acros 212918000

Code : 29

Additional Details : CAS Number : 2052-49-5 Weight : 0.66400kg Transport : UN number : 3286 Chem class : 3 Pack group : II


 View more versions of this product

Product Code. 10022740

  • 338.00 € / 800mL
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
Specifications

Specifications

Tetrabutylammonium hydroxide
2052-49-5,67-56-1
-98.0°C
[CH3(CH2)3]4NOH
0.9100g/mL
Solubility in water: soluble.
[OH-].CCCC[N+](CCCC)(CCCC)CCCC
259.48
259.46
1.3775
800 mL
1M Solutionin Methanol
65.0°C
C16H37NO
MFCD00009425
tetrabutylammonium hydroxide, tetra-n-butylammonium hydroxide, tetrabutylazanium hydroxide, tetrabutylammoniumhydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide, ammonium, tetrabutyl-, hydroxide, n,n,n-tributyl-1-butanaminium hydroxide, tetra n-butyl ammonium hydroxide, tetra-n-butyl ammonium hydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide 1:1
VDZOOKBUILJEDG-UHFFFAOYSA-M
tetrabutylazanium hydroxide
2723671
Plastic bottle
0.91
12°C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Tetrabutylammonium hydroxide, 1M solution in methanol

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity after single exposure Category 1

Hazard Statement
  • H370-Causes damage to organs.
  • H314-Causes severe skin burns and eye damage.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H225-Highly flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only