Learn More
Tetrachloroethylene, Technical, Fisher Chemical™
Brand: Fisher Chemical T/0550/17
Code : 21
Additional Details : CAS Number : 127-18-4 Weight : 4.05000kg Transport : UN number : 1897 Chem class : 6.1 Pack group : III
Packaging | Amber glass bottle |
---|---|
Quantity | 2.5L |
Chemical Identifiers
127-18-4 | |
165.82 | |
CYTYCFOTNPOANT-UHFFFAOYSA-N | |
31373 | |
1,1,2,2-tetrachloroethene |
C2Cl4 | |
MFCD00000834 | |
tetrachloroethylene, tetrachloroethene, perchloroethylene, perchlorethylene, perc, ethene, tetrachloro, tetrachlorethylene, ethylene tetrachloride, carbon dichloride, ankilostin | |
CHEBI:17300 | |
ClC(Cl)=C(Cl)Cl |
Specifications
127-18-4 | |
-22°C | |
C2Cl4 | |
Amber glass bottle | |
2.5L | |
ClC(Cl)=C(Cl)Cl | |
165.82 | |
CHEBI:17300 | |
Liquid |
Colorless | |
120°C | |
MFCD00000834 | |
tetrachloroethylene, tetrachloroethene, perchloroethylene, perchlorethylene, perc, ethene, tetrachloro, tetrachlorethylene, ethylene tetrachloride, carbon dichloride, ankilostin | |
CYTYCFOTNPOANT-UHFFFAOYSA-N | |
1,1,2,2-tetrachloroethene | |
31373 | |
165.83g/mol | |
18mbar at 20°C |
Safety and Handling
- Tetrachloroethylene
Signal Word
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H351-Suspected of causing cancer.
- H411-Toxic to aquatic life with long lasting effects.
- H336-May cause drowsiness or dizziness.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.