missing translation for 'onlineSavingsMsg'
Learn More

Tetracyanoethylene, 98%, Thermo Scientific Chemicals

Catalog Number 11935611
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
670-54-2
C6N4
128.09
NLDYACGHTUPAQU-UHFFFAOYSA-N
tetracyanoethylene, tetracyanoethene, ethylenetetracarbonitrile, tcne, ethenetetracarbonitrile, ethene, tetracyano, 1,1,2,2-ethenetetracarbonitrile, tetrakyanethylen, 1,1,2,2-tetracyanoethene, 1,1,2,2-tetracyanoethylene
12635
ethene-1,1,2,2-tetracarbonitrile
C(#N)C(=C(C#N)C#N)C#N
This item is not returnable. View return policy
670-54-2
C6N4
128.09
NLDYACGHTUPAQU-UHFFFAOYSA-N
tetracyanoethylene, tetracyanoethene, ethylenetetracarbonitrile, tcne, ethenetetracarbonitrile, ethene, tetracyano, 1,1,2,2-ethenetetracarbonitrile, tetrakyanethylen, 1,1,2,2-tetracyanoethene, 1,1,2,2-tetracyanoethylene
12635
ethene-1,1,2,2-tetracarbonitrile
C(#N)C(=C(C#N)C#N)C#N

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent, Oxidation reagent
TRUSTED_SUSTAINABILITY
Melting Point 196.0°C to 201.0°C
Boiling Point 223.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Ex N)
Packaging Glass bottle
Linear Formula (NC)2C=C(CN)2
Quantity 100 g
Fieser 01,1133; 02,397; 05,647; 06,567; 13,289
Merck Index 15,934
Solubility Information Solubility in water: hydrolyzes (forming hcn) in water. Other solubilities: slightly soluble in: ether, benzene,, carbon tetrachloride, soluble in: acetone
Formula Weight 128.09
Percent Purity 98%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material Tetracyanoethylene
Show More Show Less
compliance-icons
Product Identifier
  • Tetracyanoethylene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H310-Fatal in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'