missing translation for 'onlineSavingsMsg'
Learn More

Tetradecane, 99%, Thermo Scientific Chemicals

Catalog Number p-4510597
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
EcoSafPak™ Glass Bottle
Glass bottle
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
629-59-4
BGHCVCJVXZWKCC-UHFFFAOYSA-N
n-tetradecane, alkanes, c14-16, tridecane, methyl, ccris 715, tetradecane, analytical standard, alkanes, c14-30, myristyl, tetradekan, n-teradecane, olefine
12389
CHEBI:41253
tetradecane
CCCCCCCCCCCCCC
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
629-59-4
BGHCVCJVXZWKCC-UHFFFAOYSA-N
n-tetradecane, alkanes, c14-16, tridecane, methyl, ccris 715, tetradecane, analytical standard, alkanes, c14-30, myristyl, tetradekan, n-teradecane, olefine
12389
CHEBI:41253
tetradecane
CCCCCCCCCCCCCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 5.8°C
Color Colorless
Density 0.7670g/mL
Boiling Point 252.0°C to 254.0°C
Flash Point 99°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C14H30
Linear Formula CH3(CH2)12CH3
Refractive Index 1.4280 to 1.4300
Quantity 100 mL
Beilstein 01, 171
Specific Gravity 0.767
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol
Molecular Weight (g/mol) 198.39
Formula Weight 198.39
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Tetradecane
Show More Show Less
compliance-icons
Product Identifier
  • Tetradecane
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'