missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydro-2H-pyran-4-carbonyl chloride, ≥97%, Thermo Scientific™

Catalog Number 10423682
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
40191-32-0
C6H9ClO2
148.59
MFCD06200863
RYGUCYSSMOFTSH-UHFFFAOYSA-N
tetrahydro-2h-pyran-4-carbonyl chloride, 2h-pyran-4-carbonyl chloride, tetrahydro, tetrahydro-2h-pyran-4-carbonylchloride, pubchem12437, ksc235m5r, tetrahydropyran-4-carbonyl chloride, tetrahydro-pyran-4-carbonyl chloride, 4-chloroformyl tetrahydro-2h-pyran, tetrahydropyran-4-carboxylic chloride, 4-chlorocarbonyl tetrahydro-2h-pyran
2795505
oxane-4-carbonyl chloride
ClC(=O)C1CCOCC1
This item is not returnable. View return policy
40191-32-0
C6H9ClO2
148.59
MFCD06200863
RYGUCYSSMOFTSH-UHFFFAOYSA-N
tetrahydro-2h-pyran-4-carbonyl chloride, 2h-pyran-4-carbonyl chloride, tetrahydro, tetrahydro-2h-pyran-4-carbonylchloride, pubchem12437, ksc235m5r, tetrahydropyran-4-carbonyl chloride, tetrahydro-pyran-4-carbonyl chloride, 4-chloroformyl tetrahydro-2h-pyran, tetrahydropyran-4-carboxylic chloride, 4-chlorocarbonyl tetrahydro-2h-pyran
2795505
oxane-4-carbonyl chloride
ClC(=O)C1CCOCC1
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 148.59
Percent Purity 95%
Chemical Name or Material Tetrahydro-2H-pyran-4-carbonyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'