Learn More
tetrahydrothiopyran-4-carbonitrile, 97%, Maybridge™
Brand: Maybridge CC61916DA
Code : 31
Additional Details : CAS Number : 195503-40-3 Weight : 0.05100kg
Packaging | Amber Glass Bottle |
---|---|
Quantity | 1g |
Chemical Identifiers
195503-40-3 | |
127.205 | |
tetrahydro-2h-thiopyran-4-carbonitrile, tetrahydrothiopyran-4-carbonitrile, 2h-thiopyran-4-carbonitrile,tetrahydro, 2h-thiopyran-4-carbonitrile, tetrahydro, 4-cyanothiane, 4-cyanothian-4-yl, tetrahydro-thiopyran-4-carbonitrile | |
thiane-4-carbonitrile |
C6H9NS | |
DQKVZWAEWJUKAX-UHFFFAOYSA-N | |
21598641 | |
C1CSCCC1C#N |
Specifications
Tetrahydrothiopyran-4-carbonitrile | |
1g | |
97% | |
tetrahydro-2h-thiopyran-4-carbonitrile, tetrahydrothiopyran-4-carbonitrile, 2h-thiopyran-4-carbonitrile,tetrahydro, 2h-thiopyran-4-carbonitrile, tetrahydro, 4-cyanothiane, 4-cyanothian-4-yl, tetrahydro-thiopyran-4-carbonitrile | |
C1CSCCC1C#N | |
127.205 | |
127.21 |
Amber Glass Bottle | |
195503-40-3 | |
C6H9NS | |
DQKVZWAEWJUKAX-UHFFFAOYSA-N | |
thiane-4-carbonitrile | |
21598641 | |
97% |