missing translation for 'onlineSavingsMsg'
Learn More

Tetraisobutyldialuminoxane, 10 wt.% solution in toluene, AcroSeal™, ACROS Organics™

Brand:  Acros Organics 435988000

Code : 29

Additional Details : CAS Number : 998-00-5 Weight : 0.74360kg Transport : UN number : 1993 Chem class : 3 Pack group : II


Packaging AcroSeal™ Glass Bottle
Quantity 800mL

 View more versions of this product

Product Code. 11955971

  • 626.00 € / 800mL

Estimated Shipment
29-04-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Lewis acid
Specifications

Chemical Identifiers

998-00-5
298.42
aluminum, tetrakis 2-methylpropyl-.mu.-oxodi, tetrakis isobutyl-mu-oxodialuminium, tetraisobutyl dialumoxane, aluminum, tetrakis 2-methylpropyl-mu-oxodi, bis diisobutylaluminum oxide, bis-diisobutylaluminum-oxide, tetraisobutyldialuminoxane solution, tetraisobutyldialuminoxane solution, 10 wt. % in toluene, bis 2-methylpropyl alumanyl oxy bis 2-methylpropyl alumane
bis(2-methylpropyl)alumanyloxy-bis(2-methylpropyl)alumane
C16H36Al2O
CTNFGBKAHAEKFE-UHFFFAOYSA-N
16684086
CC(C)C[Al](CC(C)C)O[Al](CC(C)C)CC(C)C

Specifications

Tetraisobutyldialuminoxane
-8°C
0.867
Colorless
998-00-5,108-88-3
aluminum, tetrakis 2-methylpropyl-.mu.-oxodi, tetrakis isobutyl-mu-oxodialuminium, tetraisobutyl dialumoxane, aluminum, tetrakis 2-methylpropyl-mu-oxodi, bis diisobutylaluminum oxide, bis-diisobutylaluminum-oxide, tetraisobutyldialuminoxane solution, tetraisobutyldialuminoxane solution, 10 wt. % in toluene, bis 2-methylpropyl alumanyl oxy bis 2-methylpropyl alumane
CC(C)C[Al](CC(C)C)O[Al](CC(C)C)CC(C)C
298.42
298.42
0.8670g/mL
AcroSeal™ Glass Bottle
111°C
800mL
C16H36Al2O
CTNFGBKAHAEKFE-UHFFFAOYSA-N
bis(2-methylpropyl)alumanyloxy-bis(2-methylpropyl)alumane
16684086
Solution
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Tetraisobutyldialuminoxane, 10 wt.% solution in toluene

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H361d-Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P331-Do NOT induce vomiting.

SDS Documents