missing translation for 'onlineSavingsMsg'
Learn More

Tetrapropylammonium bromide, 98%, Thermo Scientific Chemicals

Catalog Number 10266790
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
1941-30-6
C12H28BrN
266.26
MFCD00011840
BGQMOFGZRJUORO-UHFFFAOYSA-M
tetrapropylammonium bromide, tetra-n-propylammonium bromide, 1-propanaminium, n,n,n-tripropyl-, bromide, tetrapropylazanium bromide, tpabr, n,n,n-tripropyl-1-propanaminium bromide, n,n,n-tripropylpropan-1-aminium bromide, tetrapropylammonium bromide tpabr, 1-propanaminium, n,n,n-tripropyl-, bromide 1:1, tetra propyl ammonium bromide
74745
CHEBI:55318
tetrapropylazanium;bromide
CCC[N+](CCC)(CCC)CCC.[Br-]
This item is not returnable. View return policy
1941-30-6
C12H28BrN
266.26
MFCD00011840
BGQMOFGZRJUORO-UHFFFAOYSA-M
tetrapropylammonium bromide, tetra-n-propylammonium bromide, 1-propanaminium, n,n,n-tripropyl-, bromide, tetrapropylazanium bromide, tpabr, n,n,n-tripropyl-1-propanaminium bromide, n,n,n-tripropylpropan-1-aminium bromide, tetrapropylammonium bromide tpabr, 1-propanaminium, n,n,n-tripropyl-, bromide 1:1, tetra propyl ammonium bromide
74745
CHEBI:55318
tetrapropylazanium;bromide
CCC[N+](CCC)(CCC)CCC.[Br-]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY
Melting Point 266°C to 272°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5 to 102.5% (Argentometry)
Packaging Plastic bottle
Linear Formula (CH3CH2CH2)4NBr
Quantity 25 g
Beilstein 04,I,364
Solubility Information Solubility in water: 100g/L water
Formula Weight 266.26
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Tetrapropylammonium bromide, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Tetrapropylammonium bromide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'