missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva (formerly Pall Lab) TF PTFE Membrane Disc Filters
Strong, chemically resistant membranes for air monitoring and sampling in aggressive environments.
362.00 € - 2440.00 €
Product Code | Brand | Pore Size | Diameter (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Diameter (Metric) | Price | Quantity & Availability | ||||
17174541
|
Cytiva (formerly Pall Lab)
66147 |
0.45 μm | 13 mm | N/A | |||||
17184541
|
Cytiva (formerly Pall Lab)
66148 |
0.45 μm | 25 mm | N/A | |||||
17144541
|
Cytiva (formerly Pall Lab)
66143 |
0.2 μm | 47 mm | N/A | |||||
17194541
|
Cytiva (formerly Pall Lab)
66149 |
0.45 μm | 47 mm | N/A | |||||
17154541
|
Cytiva (formerly Pall Lab)
66145 |
0.2 μm | 142 mm | N/A | |||||
17104551
|
Cytiva (formerly Pall Lab)
66151 |
0.45 μm | 142 mm | N/A | |||||
17114551
|
Cytiva (formerly Pall Lab)
66154 |
1 μm | 25 mm |
454.00 €
Pack of 100 |
Estimated Shipment: 23-10-2025
Log in to see stock. |
||||
17124551
|
Cytiva (formerly Pall Lab)
66155 |
1 μm | 47 mm | N/A | |||||
17134541
|
Cytiva (formerly Pall Lab)
66142 |
0.2 μm | 25 mm | N/A | |||||
17134551
|
Cytiva (formerly Pall Lab)
66159 |
1 μm | 37 mm | N/A | |||||
17164541
|
Cytiva (formerly Pall Lab)
66146 |
0.2 μm | 293 mm | N/A | |||||
Description
- Low chemical background permits highly sensitive, interference-free determinations
- Ensures accurate gravimetric determinations with low tare mass
- Ideal for filtration of gas and/or organic solvents
- This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
PTFE | |
Non-sterile |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title