Learn More
Thermo Scientific™ Thermal Printer and Paper for Eutech™ Meters
Thermal Printer for Eutech™ meters provides the convenience of printing directly from your meter whenever, wherever.
Brand: Thermo Scientific™ 01X594401
Additional Details : Weight : 0.20000kg
Description
Thermal Printer for Eutech™ meters provides the convenience of printing directly from your meter whenever, wherever.This compact thermal printer connects easily to your 110 series handheld, 450 series waterproof handheld or 2700 series benchtop meters via RS232 – ideal for data-recording and report-generation. Connecting cables included.
Features:
- Thermal
- Includes Paper-roll
- Model with real-time clock and RS232 adapter available
Specifications
Replacement paper for thermal printer, two rolls | |
Replacement Part |