Learn More
Thermo Scientific™ BioBasic™ Phenyl Drop-in Guard Cartridges
Offer convenience, economy and effective protection for extending column lifetime
Brand: Thermo Scientific™ 72405-011001
Code : 50
Additional Details : Weight : 0.10000kg
Requires : Thermo Scientific UNIGUARD™ direct connection or stand alone holder
Description
- The 10mm design offers maximum protection with minimal increase in retention
- Fit Thermo Scientific UNIGUARD direct connection and stand alone holders
- Available in packs of 4
Features of BioBasic Phenyl Solid Phase
- 300Å pore size for maximum performance with biomolecules
- Lower carbon load for optimal retention of larger peptides and proteins
- Offers different selectivity to alkyl chain chemistires for critical separations
- Excellent for LC/MS separations
Order Info
Thermo Scientific UNIGUARD™ direct connection or stand alone holder
Specifications
5 μm | |
1 mm | |
BioBasic |
10 mm | |
Phenyl |