missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BioBasic™ SCX Javelin™ Guard Columns

Offer convenient, easy to use protection for analytical HPLC columns

Brand:  Thermo Scientific™ 73205-012106

Code : A0

Additional Details : Weight : 0.10000kg

Product Code. 10547365

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • Direct-connection, fingertight design for convenience and maximum efficiency
  • Patented design requires no holder, allowing quick and easy replacement

Features of BioBasic SCX Solid Phase

  • Strong cation exchange phase based on sulfonic acid chemistry
  • Separation and retention of basic and other cationic species
  • 300Å pore size for enhanced protein and peptide separations
  • Outstanding stability under demanding pH conditions

Specifications

Specifications

5 μm
2.1 mm
BioBasic
10 mm
SCX
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers