Learn More
Thermo Scientific™ Chromacol™ Storage Vials
For general storage applications
Brand:  Thermo Scientific™ 5.3-SV
Code : 17
Additional Details : Weight : 1.26000kg
Product Code. 11961583
  • 63.62 € / Pack of 187
In Stock
to see stock.
Add to basket
Description
Description
  • Use caps with standard GPI threads
  • Class I hydrolytic extraction neutral borosilicate glass
  • May be used with injection caps with piercable seals, or solid PTFE lined storage caps
Specifications
Specifications
Clear
Borosilicate Glass
Custom External
Screw Cap
15 mm
5.0 mL
1.35 dr.
48 mm