Learn More
Thermo Scientific™ CryoMed™ Bag Freezing Presses
CryoMed™ Bag Freezing Presses
Brand:  Thermo Scientific™ 4000314
Code : NEW
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10556074
  • 871.00 € / Each
Estimated Shipment: 09-08-2024
to see stock.
Add to basket
Description
Description
CryoMed™ Bag Freezing Presses
Bag presses for Nexell, Fenwal, Gambro and Delmed. Bag capacities 2-4 bags.
Specifications
Specifications
Freezing Press
CryoMed; Nexell R4R9953 (Fenwal 4R5461)
2 bags