Learn More
Thermo Scientific™ Heat Seals for ALPS™ Microplate Heat Sealing Instruments
Save time and costs while improving sealing integrity with these microplate heat sealing instruments. Offers the capability of retrieving samples and then re-sealing the plate.
Brand: Thermo Scientific™ AB-3739
1737.56 EUR valid until 2024-12-31
Use promo code "21902" to get your promotional price.
Code : 04
Additional Details : Weight : 0.01000kg
Description
Save time and costs while improving sealing integrity with Thermo Scientific™ Heat Seals for ALPS™ Microplate Heat Sealing Instruments. Ideal for storage applications, heat sealing offers the capability of retrieving samples and then re-sealing the plate. When heat sealing, the sealing material is first placed in position over the plate, which must have a raised rim around each well. Heat is applied evenly for several seconds, resulting in binding of the sealing material to the raised rim to achieve a complete seal.
Features:
- Quick operation
- Safe for temperature-sensitive samples
- Designed for reliable performance with Thermo Scientific ALPS instruments, including the, ALPS 3000, ALPS 25 and ALPS 50 V
Specifications
Foil | |
Easy-Peel Foil Laminate, Roll, 78 mm x 610 m | |
Non-sterile | |
96 | |
1 | |
Ideal for long term storage at low temperatures; PCR | |
ALPS 5000 & ALPS 3000 | |
Abgene | |
No | |
-200°C to +90°C | |
Polypropylene, Polyethylene, Cyclic Olefin Copolymer | |
Yes |
Life Science Reagents Resistant, DMSO Resistant | |
Aluminum | |
Not Autoclavable | |
1 | |
610 m x 78 mm | |
General; Ideal for long term storage at low temperatures | |
Automation Heat Seal | |
Polypropylene, Forms weld seal with polyethylene plates, seal can be applied, removed and a new seal applied several times | |
No | |
All well shapes | |
General |