Learn More
Thermo Scientific™ Heat Seals for ALPS™ Microplate Heat Sealing Instruments
Save time and costs while improving sealing integrity with these microplate heat sealing instruments. Offers the capability of retrieving samples and then re-sealing the plate.
Brand: Thermo Scientific™ AB-3738
1649.32 EUR valid until 2024-12-31
Use promo code "21898" to get your promotional price.
Code : LE
Additional Details : Weight : 4.81950kg
Description
Save time and costs while improving sealing integrity with Thermo Scientific™ Heat Seals for ALPS™ Microplate Heat Sealing Instruments. Ideal for storage applications, heat sealing offers the capability of retrieving samples and then re-sealing the plate. When heat sealing, the sealing material is first placed in position over the plate, which must have a raised rim around each well. Heat is applied evenly for several seconds, resulting in binding of the sealing material to the raised rim to achieve a complete seal.
Features:
- Quick operation
- Safe for temperature-sensitive samples
- Designed for reliable performance with Thermo Scientific ALPS instruments, including the, ALPS 3000, ALPS 25 and ALPS 50 V
Specifications
Foil | |
Easy Pierce Foil, Roll, 78 mm x 610 m | |
Non-sterile | |
96 | |
1 | |
ideal for compound management and high throughput screening applications, suitable for PCR | |
ALPS 5000 & ALPS 3000 | |
Abgene | |
No | |
-80°C to +120°C | |
Polypropylene, Polyethylene | |
Yes |
Life Science Reagents Resistant, DMSO Resistant | |
Aluminum | |
Not Autoclavable | |
1 | |
610 m x 78 mm | |
General; Long term | |
Automation Heat Seal | |
Polypropylene | |
Yes | |
All well shapes | |
General |