Learn More
Thermo Scientific™ Evolution 300 and 600 Spectrophotometer Smart Sipper System
Easy-to-use sampling accessory featuring a reliable multi-roller peristaltic pump
Brand: Thermo Scientific™ 401322844561
Code : 04
Additional Details : Weight : 0.04500kg
Description
- For use with Thermo Scientific™ Evolution™ 300 and 600 Spectrophotometers
- Complete and automated integration into VISION and Local Control software methods
- Automatic recognition by software, smart alignment and serial number reporting
- Handles sample volumes from 200μL to 9.999mL with customizable sip volume and air gap
- Methods can be automated for sample measurement after they are sipped
Specifications
Silicone Pump Tubing | |
Evolution™ 300 and 600 spectrophotometer Smart Sipper System |