Learn More
Thermo Scientific™ Lysis Buffer for MagJET Blood gDNA Kit
Generate sample lysate in order to isolate genomic DNA. Thermo Scientific™ MagJET™ Lysate Buffer is a component of the MagJET™ Whole Blood gDNA Kits (K2741/2742) and may be purchased separately when additional solution is needed. It is provided in 15mL bottles.
Brand: Thermo Scientific™ R2341
Code : NEW
Additional Details : Weight : 0.03000kg
Description
The Thermo Scientific™ MagJET™ Whole Blood Genomic DNA Kit is designed for fast and efficient purification of genomic DNA from fresh or frozen whole blood treated with EDTA or citrate, as well as blood fractions. The kit utilizes paramagnetic bead technology, enabling high yields and robust performance. High binding capacity, uniform particle size, and rapid magnetic response of MagJET magnetic beads makes the technology ideal for high throughput automatic nucleic acid purification, as well as for manual purification by low sample throughput users. The resulting high-quality DNA is free of proteins, nucleases and other contaminants or inhibitors.
Specifications
Lysis Buffer | |
MagJET Blood gDNA Kit | |
MagJET | |
15 mL |
For Research Use Only. Not for use in diagnostic procedures.