Learn More
Thermo Scientific™ Luminaris Color HRM qPCR Master Mix
Luminaris Color HRM Master Mix is a convenient ready-to-use master mix designed for accurate detection of DNA sequence variations using High Resolution Melt analysis method.
Brand: Thermo Scientific™ K1031BID
Code : NEW
Additional Details : Weight : 0.09000kg
Description
- High performance on most real-time instruments with HRM capabilities
- Dual-color system for easy reaction setup
- Superior discrimination of all four SNP classes
- High sensitivity - accurate results from 30 copies of genomic DNA
- Exceptional stability of fully assembled reaction - for high-throughput applications
- Optimized for PikoReal Real-Time System and software
Spezifikation
Contains: • 2 x 1.25 mL Luminaris Color HRM Master Mix (2X) containing chemically modified Hot Start Taq DNA Polymerase, EvaGreen™ fluorescent dye, dNTPs and MgCl2 in an optimized PCR buffer with a blue dye; sufficient for 250 x 20 μL reactions • 1 x 1.25 mL 40X Yellow Sample Buffer • 2 x 1.25 mL Nuclease-Free Water Store at -20°C. |
|
High | |
Taq DNA Polymerase | |
SNPs (Unknown or Numerous), Epigenetic Targets | |
qPCR Master Mix | |
2X | |
250 Reactions |
HRM | |
qPCR | |
HRM (High Resolution Melting) Analysis, Mutation Scanning | |
Luminaris | |
250 Reactions | |
2 X |
For Research Use Only. Not for use in diagnostic procedures.