Learn More
Thermo Scientific™ Phire Tissue Direct PCR Master Mix
Amplify DNA directly from variety of human and animal samples with no prior purification with Phire Tissue Direct PCR Master Mix.
Brand: Thermo Scientific™ F170S
186.79 EUR valid until 2024-03-30
Use promo code "21741" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.10000kg
Description
The Phire Tissue Direct PCR Master Mix has been developed for amplification of DNA directly from a wide variety of tissues obtained from mice, human, fish, birds and insects. The Master Mix containing Phire Hot Start II DNA Polymerase is specially formulated to perform PCR in the presence of different animal tissue-derived inhibitors such as collagen, melanin and eumelanin (hair, skin) or myoglobin (muscle). The kit also includes Thermo Scientific DNARelease Additive, which can be used to improve the release of DNA from difficult tissues.
Highlights
•Direct PCR – sample is added directly to PCR reaction, therefore therese is no need for time-consuming and expensive DNA purification steps
•Dilution & Storage protocol available – allows tens of PCR reactions from one tiny sample and allows re-testing
•Specially engineered Phire DNA polymerase – extremely short PCR protocol times
•Master Mix format with premixed gel loading dye – minimizes possibility of cross-contamination, reduces sample handling and allows direct loading on gel
Applications
• Genotyping
• Transgene detection
• Knockout analysis
• Sequencing
Includes
• 2X Phire Tissue Direct PCR Master Mix
•Reaction voume with dilution & storage protocol: 20 μL
•Reaction voume with direct protocol: 50 μL
• Dilution Buffer
• DNARelease Additive
• Universal control primer mix (25 μM each)
• Water, nuclease- free
• O'GeneRuler Express DNA Ladder
Specifications
Contains: • 2 x 1.25 mL Phire Tissue Direct PCR Master Mix (2X) • 5 mL Dilution Buffer • 3 x 100 μL DNARelease Additive • 40 μL Control Primer Mix (25 μM each) • 2 x 1.25 mL Water, nuclease- free • 50 μg O'GeneRuler Express DNA Ladder (sufficient for 100 applications) Store at -20°C. |
|
Direct PCR | |
Fast | |
Phire | |
250 x 20 μL Reactions | |
Genotyping | |
250 x 20 mL Reactions | |
SuperMix or Master Mix | |
DNA |
Medium | |
Phire Hot Start II DNA polymerase | |
Yes | |
Direct PCR Master Mix | |
250 x 20 μL Reactions | |
Built-In Hot Start | |
Blunt | |
Tissue |
Safety and Handling
- Phire Tissue Direct PCR Master Mix/DNARelease Additive
Signal Word
- Danger
Hazard Category
- Respiratory sensitiser Category 1
Hazard Statement
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P501-Dispose of contents/container to .
Supplemental information
- MIXTURE LIST-Contains : Proteinase, tritirachium album serine
For Research Use Only. Not for use in diagnostic procedures.