missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ General-Purpose Coverslips

Brand:  Epredia™ 1001624

Code : A0

Additional Details : Weight : 0.01000kg

Product Code. 12580047

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • Free from blisters and flaws
  • Cleaned and polished
  • Consistent in cut, thickness, and dimensional accuracy

Specifications

Specifications

Coverslip
Glass
70 mm
Ideal for use on the Shandon Consul™ automated coverslipper
Rectangle
22 mm
1 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers