Learn More
Thermo Scientific™ H-6000A Bucket Adapters
Accept a variety of blood processing tubes, bags and bottles
Brand: Thermo Scientific™ 00512
Code : NEW
Additional Details : Weight : 1.00000kg
Description
Accept a variety of blood processing tubes, bags and bottles
For use with Thermo Scientific H-6000A buckets.
- Adapters hold a maximum capacity of 6 × 500mL (bottles), 6 × 550mL (blood bags), and 132 × 7mL (tubes). Max. O.D. × L: 70 x 160mm
Specifications
6 x 550 mL Blood Bags | |
H-6000A bucket for Thermo Scientific™ Sorvall™ RC3BP™ Plus low speed centrifuge rotor |