Learn More
Thermo Scientific™ Hypersil™ ODS (C18) Javelin™ Guard Columns
Offer convenient, easy-to-use protection for analytical HPLC columns
416.39 €
Product Code | Brand | Diameter (Metric) Inner | Length (Metric) | Particle Size | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inner | Length (Metric) | Particle Size | Price | Quantity & Availability | |||
11533181
|
Thermo Scientific™
30105-024006 |
4 mm | 20 mm | 5 μm |
416.39 €
Pack of 4 |
Estimated Shipment: 02-05-2024
Log in to see stock. |
|||
11503181
|
Thermo Scientific™
30105-014006 |
4 mm | 10 mm | 5 μm |
416.39 €
Pack of 4 |
Estimated Shipment: 02-05-2024
Log in to see stock. |
|||
11523181
|
Thermo Scientific™
30105-022106 |
2.1 mm | 20 mm | 5 μm |
416.39 €
Pack of 4 |
Estimated Shipment: 02-05-2024
Log in to see stock. |
|||
Description
- Direct-connection, fingertight design for convenience and maximum efficiency
- Patented design requires no holder, allowing quick and easy replacement
Specifications
C18 |