Learn More
Epredia™ Wright-Giemsa Stain
Produces somewhat darker results when compared with Giemsa, particularly in the blue to magenta shades
Brand: Epredia™ 89013
Code : A0
Additional Details : Weight : 0.01000kg Transport : UN number : 1230 Chem class : 3 Pack group : II
Description
Produces somewhat darker results compared with Giemsa, particularly in the blue to magenta shades.
Specifications
32 oz. | |
Liquid | |
Produces somewhat darker results compared with giemsa, particularly in the blue to magenta shades | |
-94°C |
65°C | |
Wright-Giemsa Stain | |
Purple | |
Bottle |