Learn More
Thermo Scientific™ ABgene™ EasyStrip PCR Tubes
Each tube in the strip has an individually sealable cap to reduce the chance of sample contamination. PCR and QPCR setup and analysis are made much simpler with the unique design of these new tubes.
265.00 € - 292.00 €
Specifications
Quantity 250 Strips
For Use With (Equipment) 0.2mL Cycler Blocks
View More Specs
Products 3
Product Code Brand Closure Type Color Description Price Quantity & Availability  
Product Code Brand Closure Type Color Description Price Quantity & Availability  
12154711
View Documents Promotion Details
Thermo Scientific™
AB1502
Ultra-clear Flat Cap Clear For PCR or QPCR This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12164711
View Documents Promotion Details
Thermo Scientific™
AB1502W
Ultra-clear Flat Cap White For QPCR This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10656154
View Documents Promotion Details
Thermo Scientific™
AB1504
Domed Clear For PCR This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
For QPCR, EasyStrip Snap Tubes feature Ultra Clear Caps tethered to either white or natural tubes. White wells have been proven to increase assay sensitivity and reproducibility. The attached Ultra Clear Cap is an optically clear window into the reaction effectively allowing fluorescent signal to pass through unaffected, facilitating the benefits of white plastic.
In standard PCR, domed caps are used to allow for the best seal when using thermal cyclers with a screw down lid. The domed capped tubes are available in natural. With attached caps, pre- and post- aliquoting steps are much easier to perform. They allow individual access to each tube without compromising any steps in your workflow.
  • 8-strip PCR tubes with individually attached caps
  • Each cap can be opened and closed separately, preventing cross-contamination
  • The only strip tubes available that combine optimal conditions for QPCR detectiondash (em)white tubes and ultra clear attached caps
  • Individually lettered caps help to orient the tubes and aid in verifying sample identity
  • Tubes feature uniform, thin walls and are certified free from RNase, DNase, endotoxins and contaminating DNAbullets1 (end)
Specifications
Specifications
250 Strips
0.2mL Cycler Blocks