missing translation for 'onlineSavingsMsg'
Learn More

Hanna Instruments™ Thermocouple Probe Hi-766Pb Interchangeable K Type For Hot Solids

Brand:  Hanna Instruments™ HI766PB

Code : 03

Additional Details : Weight : 0.01000kg

Product Code. 10448452

  • 188.00 € / Each
Estimated Shipment: 10-05-2024
to see stock.
Add to basket

Description

Description

Furnaces and moulds stainless steel surface 650°C max 200mm x 16mm diameter Hanna
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers