missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HUBER Thermofluid DW-Therm HT
Click to view available options
Quantity:
10 L
Unit Size:
10 L
Description
- Thermofluid DW-Therm HT P20.340.32
- Unistat-thermofluid for closed systems
- Min. temp: 20°C
- Max. temp.: (170)340°C
- Viscosity at 25°C: 32 mm2/sec
Specifications
Specifications
| Color | Orange |
| Temperature Range | 20°C to 340°C |
| Type | Thermofluid DW-Therm HT |
| Quantity | 10 L |
| Temperature (Metric) Maximum | 340°C |
| Viscosity | 32 mm2/sec. |
Safety and Handling
Product Identifier
- DW-Therm HT
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction