missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermoplastic Elastomer Ear Plug
missing translation for 'orderingAttributeHoverText'
Size:
Universal
Specifications
Color | Orange |
Certifications/Compliance | EC 030649/ 040809/ 040950, ISO 9001/2000 |
Size | Universal |
For Use With (Application) | For comfortable, personalized fit |
Quantity | 50/Pk. |
Plug Shape | Flanged |
Product Type | Ear Plug |
Disposable | Single-use |
Material | TPE |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'