Learn More
Thiamphenicol, Thermo Scientific™
The methyl-sulfonyl analogue of chloramphenicol
32.40 € - 265.00 €
Chemical Identifiers
| CAS | 15318-45-3 |
|---|---|
| Molecular Formula | C12H15Cl2NO5S |
| Molecular Weight (g/mol) | 356.214 |
| MDL Number | MFCD00467983 |
| InChI Key | OTVAEFIXJLOWRX-NXEZZACHSA-N |
| Synonym | thiamphenicol, thiophenicol, thiamcol, +-thiamphenicol, raceophenidol, thiocymetin, dextrosulphenidol, thiamphenicolum, d-thiocymetin, d-thiophenicol |
| PubChem CID | 27200 |
| ChEBI | CHEBI:32215 |
| IUPAC Name | 2,2-dichloro-N-[(1R,2R)-1,3-dihydroxy-1-(4-methylsulfonylphenyl)propan-2-yl]acetamide |
| SMILES | CS(=O)(=O)C1=CC=C(C=C1)C(C(CO)NC(=O)C(Cl)Cl)O |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15485769
|
Thermo Scientific Alfa Aesar
J63575.03 |
1g |
32.40 €
1g |
In Stock
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
|
15495769
|
Thermo Scientific Alfa Aesar
J63575.06 |
5g |
66.20 €
5g |
In Stock
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
|
15405779
|
Thermo Scientific Alfa Aesar
J63575.14 |
25g |
265.00 €
25g |
Estimated Shipment: 23-09-2026
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsThiamphenicol is used to treat chancroid in men and uncomplicated gonorrhea. It is used in studies of bacterial protein synthesis at the level of peptidyl transferase activity associated with the 23S rRNA of the 50S ribosomal subunit. It is used to study chloraniphenicol-thiamphenicol-resistance and the use of fluorinated analogs when resistance is encountered. Its main advantage over chloramphenicol is that it has never been associated with aplastic anaemia.
Solubility
Soluble in acetonitrile or DMF. Slightly soluble in water
Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
Chemical Identifiers
| 15318-45-3 | |
| 356.214 | |
| OTVAEFIXJLOWRX-NXEZZACHSA-N | |
| 27200 | |
| 2,2-dichloro-N-[(1R,2R)-1,3-dihydroxy-1-(4-methylsulfonylphenyl)propan-2-yl]acetamide |
| C12H15Cl2NO5S | |
| MFCD00467983 | |
| thiamphenicol, thiophenicol, thiamcol, +-thiamphenicol, raceophenidol, thiocymetin, dextrosulphenidol, thiamphenicolum, d-thiocymetin, d-thiophenicol | |
| CHEBI:32215 | |
| CS(=O)(=O)C1=CC=C(C=C1)C(C(CO)NC(=O)C(Cl)Cl)O |
Specifications
| 15318-45-3 | |
| MFCD00467983 | |
| 14,9301 | |
| Soluble in acetonitrile or DMF; Slightly soluble in water | |
| CS(=O)(=O)C1=CC=C(C=C1)C(C(CO)NC(=O)C(Cl)Cl)O | |
| 356.214 | |
| CHEBI:32215 | |
| Thiamphenicol |
| C12H15Cl2NO5S | |
| 2819542 | |
| thiamphenicol, thiophenicol, thiamcol, +-thiamphenicol, raceophenidol, thiocymetin, dextrosulphenidol, thiamphenicolum, d-thiocymetin, d-thiophenicol | |
| OTVAEFIXJLOWRX-NXEZZACHSA-N | |
| 2,2-dichloro-N-[(1R,2R)-1,3-dihydroxy-1-(4-methylsulfonylphenyl)propan-2-yl]acetamide | |
| 27200 | |
| 356.22 |
Safety and Handling
missing translation for 'einecsNumber' : 239-355-3
missing translation for 'rtecsNumber' : AB6680000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.