missing translation for 'onlineSavingsMsg'
Learn More

Thionyl chloride, 99.5+%, Thermo Scientific Chemicals

Catalog Number p-200191
24.65 EUR valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
7719-09-7
MFCD00011449
24386
CHEBI:29290
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
7719-09-7
MFCD00011449
24386
CHEBI:29290

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY
Color Colorless through Yellow
Physical Form Liquid
Quantity 100 g
Assay Percent Range 99.5+%
Linear Formula SOCl2
Fieser 01,1158; 02,412; 03,290; 04,503; 05,663; 06,585; 07,366; 08,481; 09,465; 10,399; 12,204; 13,297
Formula Weight 118.97
Percent Purity ≥99.50%
Packaging Glass bottle
Density 1.6400g/mL
Chemical Name or Material Thionyl chloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Thionyl chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'