missing translation for 'onlineSavingsMsg'
Learn More

Thionyl chloride, 99.7%, ACROS Orgaincs™

Brand:  Acros Organics 382662500

Code : 29

Additional Details : CAS Number : 7719-09-7 Weight : 0.46000kg Transport : UN number : 1836 Chem class : 8 Pack group : I


Quantity 250 mL

 View more versions of this product

Product Code. 10002212

Quantity Price
1 31.12 € / 250mL
10 28.53 € / 250mL
Estimated Shipment: 03-08-2021

Please to purchase this item. Need a web account? Register with us today!



Sea freight only: For EU stock allow up to 15 days delivery.

Description

Description

Specifications

Chemical Identifiers

7719-09-7
118.96
sulfurooyl dichloride
Cl2OS
FYSNRJHAOHDILO-UHFFFAOYSA-N
ClS(Cl)=O

Specifications

Thionyl chloride
Glass bottle
1.64
76.0°C
250 mL
99.7%
SOCl2
Solubility aq. soln.: reacts. Other solubilities: miscible with benzene, chloroform and carbon, tetrachloride
ClS(Cl)=O
118.96
1.6400 g/mL
1.5190 to 1.5210
0.6 mPa.s (20°C)
-105.0°C
7719-09-7
Cl2OS
15, 9501
FYSNRJHAOHDILO-UHFFFAOYSA-N
sulfurooyl dichloride
118.97
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Thionyl chloride

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 3
  • Specific target organ toxicity Category 3

Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P402+P404-Store in a dry place. Store in a closed container.

SDS Documents
Certificates