missing translation for 'onlineSavingsMsg'
Learn More

Thiophosgene, 85%, ACROS Organics™

Brand:  Acros Organics 151640025

Code : 29

Additional Details : CAS Number : 463-71-8 Weight : 4.00000kg Transport : UN number : 2474 Chem class : 6.1 Pack group : I


Packaging Glass bottle
Quantity 2.5kg

 View more versions of this product

Product Code. 11944391

  • 1216.00 € / 2.50kg

Estimated Shipment: 15-07-2021

Please to purchase this item. Need a web account? Register with us today!


Sea freight only: For EU stock allow up to 15 days delivery.

Description

Description

C-X bond formation reagent
Specifications

Chemical Identifiers

463-71-8
114.98
ZWZVWGITAAIFPS-UHFFFAOYSA-N
10040
thiocarbonyl dichloride
CCl2S
MFCD00004918
thiophosgene, carbonothioic dichloride, carbon chlorosulfide, thiocarbonyl chloride, thiocarbonic dichloride, dichlorothiocarbonyl, thiofosgen, phosgene, thio, thiokarbonylchlorid, carbonyl chloride, thio
CHEBI:29366
C(=S)(Cl)Cl

Specifications

Thiophosgene
80% min. (GC)
ClCSCl
03,134
thiophosgene, carbonothioic dichloride, carbon chlorosulfide, thiocarbonyl chloride, thiocarbonic dichloride, dichlorothiocarbonyl, thiofosgen, phosgene, thio, thiokarbonylchlorid, carbonyl chloride, thio
ZWZVWGITAAIFPS-UHFFFAOYSA-N
thiocarbonyl dichloride
10040
114.98
85%
Glass bottle
Solubility in water: slow decomposition in cold, fast in hot water. Other solubilities: soluble in ether, ccl4, decomposes in alcohol
73.0°C
2.5kg
463-71-8
CCl2S
MFCD00004918
05,667; 09,466
Solubility in water: slow decomposition in cold, fast in hot water. Other solubilities: soluble in ether, ccl4, decomposes in alcohol
C(=S)(Cl)Cl
114.98
CHEBI:29366
Liquid
1.5080g/mL
1.5442
1.508
Orange-Red to Red
62°C
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Thiophosgene

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H330-Fatal if inhaled.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

SDS missing translation for 'documents'
Product Certifications