Learn More
Terumo™ Three-part Syringes
Use these syringes for a variety of applications. Terumo™ Three-part Syringes meet the highest quality standards, from development to packaging.
Brand:  Terumo™ 8SS05S1
Additional Details : Weight : 1.00000kg
Product Code. 15573104
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Highly transparent barrel for easy recognition of air bubbles 
  • Clear printing for precise dosage management
  • Latex-free syringes contain no natural rubber 
  • Environmental-friendly Designed to have a minimum of waste
Specifications
Specifications
Class IIa, sterile product following MDD 93/42/EEC
Clear
Yes
Yes
5 mL
Yes
Polypropylene, Silicone Oil (Lubricant), TPE
Injection of fluids - immediately after filling - into the human body, for aspiration of fluids or for the sampling of blood or other body fluids
3-part Syringe
0.2 mL
Concentric
Sterile