missing translation for 'onlineSavingsMsg'
Learn More

TIA1 Mouse, Clone: TIA1/1313, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio 7072-MSM1-P1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15990968

  • 459.10 € / 100µg

Estimated Shipment: 30-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

TIA1
Cytoplasmic
200μg/mL
10mM PBS with 0.05% BSA and 0.05% Azide
TIA1
Recombinant human TIA1 fragment of 102 amino acid residues (aa279-380)
40kDa
Protein A/G Purified
RUO
Primary
7072
Western Blot
TIA1/1313
Unconjugated
XP_016860286
Mouse
IgG2b
HepG2, PC3 and HeLa cells. Tonsil.
100 μg
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Monoclonal
Videos
SDS
Documents
Certificates