missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIM-3 Antibody (344823R), mFluor Violet 500 SE, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB2365RMFV500
555.90 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
TIM-3 Monoclonal antibody specifically detects TIM-3 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| TIM-3 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rat | |
| 0.1 mL | |
| Cancer, Immunology, Innate Immunity, Neuroscience | |
| 84868 | |
| Store at 4C in the dark. |
| Flow Cytometry, CyTOF | |
| 344823R | |
| Flow Cytometry, CyTOF-ready | |
| CD366, FLJ14428, HAVcr-2, hepatitis A virus cellular receptor 2, kidney injury molecule-3, T cell immunoglobulin mucin 3, T cell immunoglobulin mucin-3, T-cell immunoglobulin and mucin domain-containing protein 3, TIM 3, Tim-3, TIM3 T-cell membrane protein 3, TIMD-3, TIMD3KIM-3 | |
| Mouse myeloma cell line NS0-derived recombinant human TIM-3, Ser22-Arg200, Accession # Q8TDQ0.2 | |
| RUO | |
| Primary | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction