missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ Tintometer™ Bromophenol Blue Solution

For use with Lovibond test kits, colorimeters and spectrophotometers

Brand:  Lovibond™ 451622

Code : NEW

Additional Details : Weight : 0.30000kg

 View more versions of this product

Product Code. 15322025

  • 53.60 € / 250mL


Estimated Shipment
16-06-2020
Add to basket

Description

Description

  • Bromophenol Blue Solution
  • pH Indicator
  • 100mL and 250mL Bottles
Specifications

Specifications

250mL
For use with Lovibond test kits, colorimeters and spectrophotometers
pH Indicator
Bromophenol Blue Solution
pH Test
Videos
Documents