Learn More
Lovibond™ Tintometer™ CHECKIT™ Comparator, Sulphite LR Test, 0.5 to 10mg/L
Suitable for both mobile and static water analysis. Checkit™ Comparators are available with a number of different color scales and provide the basis for a comprehensive easy to to to use analysis system.
Brand:  Lovibond™ 147380
Additional Details : Weight : 0.45000kg
Product Code. 10254692
  • 63.60 € / Each
Estimated Shipment: 17-05-2024
to see stock.
Add to basket
Description
Description
  • The Lovibond™ Checkit™ comparator is a compact, lightweight colorimetric unit
  • Suitable for both mobile and static water analysis
  • Available with a number of different color scales
  • Provides the basis for a comprehensive easy to to to use analysis system
  • Each Checkit™ Comparator Disc contains a continuous colour scale for exact color matching between standard and sample
  • Each Checkit™ Comparator Kit is supplied in a carry case with comparator, vials, discs, reagents and method instructions for all tests
  • The Sulphite disc supplied measures from 0.5 to 10mg/L with the following steps: 0.5 / 1 / 1.5 / 2 / 2.5 / 3 / 3.5 / 4 / 4.5 / 5 / 6 / 7 / 8 / 9 / 10.5 / 4.0
Specifications
Specifications
Water Quality Testing
Carry case with Comparator, Vials, Discs, reagents and method instructions for all tests
Sulphite LR Test
Checkit™ Comparator
0.5 to 10mg/L