Learn More
Lovibond™ Tintometer™ DPD No. 3 High Calcium Tablets
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
24.80 € - 94.80 €
Specifications
For Use With (Application) | Photometry Tablet Reagents |
---|---|
For Use With (Equipment) | Test kits, colorimeters and spectrophotometers |
Type | DPD No. 3 High Calcium Tablets |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15301975
|
Lovibond™
515731BT |
250 Tablets |
54.70 €
Pack of 250 |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||||
15311975
|
Lovibond™
515732BT |
500 Tablets |
94.80 €
Pack of 500 |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||||
15391965
|
Lovibond™
515730BT |
100 Tablets |
24.80 €
Pack of 100 |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||||
Description
- New aluminum foil blister packaging brings added convenience to the tradition of protection achieved in Lovibond’s long established tablet production technology
- With the new blister strip, the user just pushes the tablet through the protective foil, straight into the sample
- Each tablet is contained within an individually formed foil cup, lined with the latest aluminum composite material, and guaranteeing product performance
- Blister pack has been reduced in size to 91 x 34mm for convenient storage and shipping
- ‘BT’ is added to the end of the code to identify the new style of packaging
- Simple, time-saving and practical
- There are no safety risks if the tablets are used in line with the instructions supplied
- Safety data sheets are available for all reagents
Specifications
Photometry Tablet Reagents | |
Test kits, colorimeters and spectrophotometers | |
DPD No. 3 High Calcium Tablets |