Learn More
Tintometer™ Lovibond™ Water Testing Reagents
For use in water testing
Brand:  Tintometer™ 530550
Additional Details : Weight : 0.09000kg
Product Code. 13223459
  • 50.00 € / Each
Estimated Shipment: 20-06-2024
to see stock.
Add to basket
Description
Description
  • Iron Vario TPTZ F10
  • Available as powder pack
  • For water, waste water, seawater testing
Specifications
Specifications
For water testing, waste water testing, seawater testing
100/Pk.
Iron vario TPTZ F10 reagent powder
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Vario TPTZ Iron F10Ml
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • EUH031-Contact with acids liberates toxic gas.
  • EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : disodium disulphite,sodium dithionite,methenamine