Learn More
Lovibond™ Tintometer™ Nitrate Test Powder, 100 Tests
For use with Lovibond test kits, colorimeters and spectrophotometers
Brand: Lovibond™ 465230
Code : NEW
Additional Details : Weight : 0.01500kg Transport : UN number : 3077 Chem class : 9 Pack group : III
Description
For use with Lovibond test kits, colorimeters and spectrophotometers
- Nitrate Test Powder
- 100 Tests
- For 605/705 Compatators, 942/975 Colorimeters, 694 Soil Test Kit
Specifications
605/705 Compatators, 942/975 Colorimeters, 694 Soil Test Kit | |
100 | |
Nitrate Test Powder |
Safety and Handling
- nitrate test powder
Signal Word
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
Hazard Statement
- H410-Very toxic to aquatic life with long lasting effects.
- H400-Very toxic to aquatic life.
Precautionary Statement
- P273-Avoid release to the environment.
- P391-Collect spillage.
- P501-Dispose of contents/container to in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : zinc powder - zinc dust (stabalized)