missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Titanium aluminide, 99.5% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
Titanium aluminide is used in aircraft turbine engines, exhaust valves and turbochargers for passenger vehicle engines, intermediate process for some mill products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Physical Form | Powder |
Quantity | 100 g |
UN Number | UN3178 |
Solubility Information | Insoluble in water. |
Percent Purity | 99.5% |
Packaging | Plastic bottle |
Chemical Name or Material | Titanium aluminide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'