missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Titanium(IV) isopropoxide, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 mL
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
Specifications
Melting Point | >15.0°C |
Density | 0.9500g/mL |
Boiling Point | 240.0°C |
Flash Point | 46°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 16.6 to 17.3% (Ti) |
Packaging | Glass bottle |
Linear Formula | Ti[OCH(CH3)2]4 |
Refractive Index | 1.4654 to 1.4684 |
Quantity | 5 mL |
Show More |
Safety and Handling
Product Identifier
- Titanium(IV) isopropoxide
- Danger
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H331-Toxic if inhaled.
- H336-May cause drowsiness or dizziness.
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'