missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) n-butoxide, 99+%, Thermo Scientific Chemicals

Catalog Number p-7080342
Click to view available options
Quantity:
10 g
100 g
500 g
This item is not returnable. View return policy
5593-70-4
C16H40O4Ti
344.359
MFCD00009433
FPCJKVGGYOAWIZ-UHFFFAOYSA-N
butyl titanate, butan-1-ol; titanium, titanium n-butoxide, tetrabutyl orthotitanate, tetrabutyl titanate, 1-butanol; titanium, tetra butyl orthotitanate, tetrabutyl ortho-titanate
25203927
butan-1-ol;titanium
CCCCO.CCCCO.CCCCO.CCCCO.[Ti]
This item is not returnable. View return policy
5593-70-4
C16H40O4Ti
344.359
MFCD00009433
FPCJKVGGYOAWIZ-UHFFFAOYSA-N
butyl titanate, butan-1-ol; titanium, titanium n-butoxide, tetrabutyl orthotitanate, tetrabutyl titanate, 1-butanol; titanium, tetra butyl orthotitanate, tetrabutyl ortho-titanate
25203927
butan-1-ol;titanium
CCCCO.CCCCO.CCCCO.CCCCO.[Ti]

Titanium(IV) n-butoxide is used in the preparation of nanosized titania powders in the anatase form and ferroelectric bismuth titanate thin films. It is also used as intermediates, paint additives, coating additives, processing aids and as process regulators. It is involved in the preparation of nanocrystalline titanium dioxide powders at room temperature.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 310°C to 314°C
Melting Point -55°C
Color Yellow
Physical Form Liquid
Quantity 10 g
Linear Formula Ti[O(CH2)3CH3]4
UN Number UN2924
Beilstein 4148236
Flash Point 41°C (170°F)
Solubility Information Miscible with aliphatic,aromatic,chlorinated and oxygenated solvents.
Formula Weight 340.36
Percent Purity ≥99%
Odor Alcohol-like
Refractive Index 1.49
Sensitivity Moisture sensitive
Density 0.998 g/mL
Chemical Name or Material Titanium (IV) n-butoxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'