missing translation for 'onlineSavingsMsg'
Learn More

Titanium Standard For ICP, 1000μg/mL in H2O/0.24 F, Assurance Grade, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Brand:  SPEX Certiprep PLTI9-2Y

Code : A0

Additional Details : CAS Number : 7440-32-6 Weight : 0.12700kg

Packaging Triple-leached LDPE bottle
Quantity 125mL

 View more versions of this product

Product Code. 11973743

  • 51.94 € / 125mL

Estimated Shipment
04-05-2021

Please to purchase this item. Need a web account? Register with us today!


Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Ti1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Identifiers

16962-40-6

Specifications

Titanium Standard
Matrix    Concentration    Cat. No.
H2O/tr 2.4%F-     1000μg/mL    PLTI9-2Y, -2T, -2X
40% HCl    1000μg/mL    PLTI5-2X
40% HCl    10,000μg/mL    PLTI5-3X
H2O/tr 2.4%F-    10,000μg/mL    PLTI9-3Y, -3X
16962-40-6,7732-18-5,7732-18-5
79.9
Triple-leached LDPE bottle
H2O/0.24 F
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
79.9
Assurance
Assurance Certificate of Analysis reports up to 30 elements at ppb levels.
125mL
Videos
SDS
Documents

Product intended for laboratory use only