missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNF Mouse anti-Human, Clone: P/T2, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB14453.100ug
Additional Details : Weight : 0.02000kg
Description
Mouse monoclonal antibody raised against hexadecapeptide of human TNF.
- High specificity
- High reactivity
Specifications
TNF | |
Monoclonal | |
Unconjugated | |
Flow Cytometry (0.5-1 ug/million cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (2-4 ug/mL) The optimal working dilution should be determined by the end user. | |
P01375 | |
TNF | |
A hexadecapeptide (conjugated with thyroglobulin) corresponding to amino acids 115-130 of human TNF. | |
100 μg | |
Primary | |
Human | |
Liquid |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed) | |
P/T2 | |
Mouse monoclonal antibody raised against hexadecapeptide of human TNF. | |
In 10 mM PBS (0.05% BSA and 0.05% azide). | |
DIF/TNF-alpha/TNFA/TNFSF2 | |
Mouse | |
Polyethylene Glycol (PEG) precipitation | |
RUO | |
7124 | |
Store at 4°C. | |
IgM κ |
For Research Use Only (RUO)